RIE156324634 Rieke Metals, Inc.

4-(3-pyrrolinomethyl)-4'-thiomethylbenzophenone

  • MDL number MFCD03841850
  • Cas Number : 898763-97-8
  • Molecular Weight : 309.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl][4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C19H19NOS/c1-22-18-10-8-17(9-11-18)19(21)16-6-4-15(5-7-16)14-20-12-2-3-13-20/h2-11H,12-14H2,1H3
Inchi Key KPYQJBYPYKLZKF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0900-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-15
Availability details for
Quantity
  • Availability
  • Add to Cart