RIE156324642 Rieke Metals, Inc.

2,4-dimethyl-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841858
  • Cas Number : 898764-13-1
  • Molecular Weight : 291.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-27
Availability details for
Quantity
  • Availability
  • Add to Cart