RIE156324652 Rieke Metals, Inc.

4'-(3-pyrrolinomethyl)-2-trifluoromethylbenzophenone

  • MDL number MFCD03841868
  • Cas Number : 898764-43-7
  • Molecular Weight : 331.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-43
Availability details for
Quantity
  • Availability
  • Add to Cart