RIE156324655 Rieke Metals, Inc.

4-bromo-2-fluoro-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841871
  • Cas Number : 898764-52-8
  • Molecular Weight : 360.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-49
Availability details for
Quantity
  • Availability
  • Add to Cart