RIE156324656 Rieke Metals, Inc.

2-chloro-4-fluoro-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841872
  • Cas Number : 898764-55-1
  • Molecular Weight : 315.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-50
Availability details for
Quantity
  • Availability
  • Add to Cart