RIE156324661 Rieke Metals, Inc.

2,5-dichloro-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841877
  • Cas Number : 898764-67-5
  • Molecular Weight : 332.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dichlorophenyl)[4-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C18H15Cl2NO/c19-15-7-8-17(20)16(11-15)18(22)14-5-3-13(4-6-14)12-21-9-1-2-10-21/h1-8,11H,9-10,12H2
Inchi Key ZFJYXOIBQKPAFO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0900-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-55
Availability details for
Quantity
  • Availability
  • Add to Cart