RIE156324665 Rieke Metals, Inc.

3,4-difluoro-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841881
  • Cas Number : 898764-75-5
  • Molecular Weight : 299.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-59
Availability details for
Quantity
  • Availability
  • Add to Cart