RIE156324666 Rieke Metals, Inc.

3,5-difluoro-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841882
  • Cas Number : 898764-77-7
  • Molecular Weight : 299.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-60
Availability details for
Quantity
  • Availability
  • Add to Cart