RIE156324670 Rieke Metals, Inc.

cyclopentyl 4-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03841886
  • Cas Number : 898764-85-7
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0903-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0903-00
Availability details for
Quantity
  • Availability
  • Add to Cart