RIE156324671 Rieke Metals, Inc.

cyclohexyl 4-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03841887
  • Cas Number : 898764-87-9
  • Molecular Weight : 269.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0904-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0904-00
Availability details for
Quantity
  • Availability
  • Add to Cart