RIE156324677 Rieke Metals, Inc.

3-(morpholinomethyl)benzophenone

  • MDL number MFCD03841893
  • Cas Number : 763863-61-2
  • Molecular Weight : 281.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-01
Availability details for
Quantity
  • Availability
  • Add to Cart