RIE156324681 Rieke Metals, Inc.

2-methoxy-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841897
  • Cas Number : 898765-05-4
  • Molecular Weight : 311.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-05
Availability details for
Quantity
  • Availability
  • Add to Cart