RIE156324682 Rieke Metals, Inc.

3-methoxy-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841898
  • Cas Number : 898765-07-6
  • Molecular Weight : 311.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-06
Availability details for
Quantity
  • Availability
  • Add to Cart