RIE156324683 Rieke Metals, Inc.

4'-methoxy-3-morpholinomethyl benzophenone

  • MDL number MFCD03841899
  • Cas Number : 898765-09-8
  • Molecular Weight : 311.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-07
Availability details for
Quantity
  • Availability
  • Add to Cart