RIE156324693 Rieke Metals, Inc.

4'-bromo-3-morpholinomethyl benzophenone

  • MDL number MFCD03841909
  • Cas Number : 898765-38-3
  • Molecular Weight : 360.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-19
Availability details for
Quantity
  • Availability
  • Add to Cart