RIE156324698 Rieke Metals, Inc.

2,3-dimethyl-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841914
  • Cas Number : 898765-53-2
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-26
Availability details for
Quantity
  • Availability
  • Add to Cart