RIE156324702 Rieke Metals, Inc.

3,4-dimethyl-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841918
  • Cas Number : 898791-74-7
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)[3-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO2/c1-15-6-7-19(12-16(15)2)20(22)18-5-3-4-17(13-18)14-21-8-10-23-11-9-21/h3-7,12-13H,8-11,14H2,1-2H3
Inchi Key KOAHESXSDJDPSV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0100-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-30
Availability details for
Quantity
  • Availability
  • Add to Cart