RIE156324706 Rieke Metals, Inc.

3-chloro-4-fluoro-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841922
  • Cas Number : 898791-86-1
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-4-fluorophenyl)[3-(4-morpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17ClFNO2/c19-16-11-15(4-5-17(16)20)18(22)14-3-1-2-13(10-14)12-21-6-8-23-9-7-21/h1-5,10-11H,6-9,12H2
Inchi Key OTPXHBYUEIYAPQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0100-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-34
Availability details for
Quantity
  • Availability
  • Add to Cart