RIE156324708 Rieke Metals, Inc.

2-fluoro-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841924
  • Cas Number : 898791-92-9
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-37
Availability details for
Quantity
  • Availability
  • Add to Cart