RIE156324710 Rieke Metals, Inc.

3'-morpholinomethyl-2-trifluoromethylbenzophenone

  • MDL number MFCD03841925
  • Cas Number : 898791-98-5
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-44
Availability details for
Quantity
  • Availability
  • Add to Cart