RIE156324714 Rieke Metals, Inc.

3-chloro-5-fluoro-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841930
  • Cas Number : 898792-10-4
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-51
Availability details for
Quantity
  • Availability
  • Add to Cart