RIE156324718 Rieke Metals, Inc.

2,5-dichloro-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841934
  • Cas Number : 898792-22-8
  • Molecular Weight : 350.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-55
Availability details for
Quantity
  • Availability
  • Add to Cart