RIE156324719 Rieke Metals, Inc.

3,4-dichloro-3'-morpholinomethyl benzophenone

  • MDL number MFCD03841935
  • Cas Number : 898792-24-0
  • Molecular Weight : 350.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0100-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0100-56
Availability details for
Quantity
  • Availability
  • Add to Cart