RIE156324725 Rieke Metals, Inc.

Cyclopropyl 3-(morpholinomethyl)phenyl ketone

  • MDL number MFCD03841941
  • Cas Number : 898792-36-4
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0101-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0101-00
Availability details for
Quantity
  • Availability
  • Add to Cart