RIE156324731 Rieke Metals, Inc.

ethyl 6-[3-(morpholinomethyl)phenyl]-6-oxohexanoate

  • MDL number MFCD03841947
  • Cas Number : 898792-48-8
  • Molecular Weight : 333.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0107-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0107-00
Availability details for
Quantity
  • Availability
  • Add to Cart