RIE156324732 Rieke Metals, Inc.

ethyl 7-[3-(morpholinomethyl)phenyl]-7-oxoheptanoate

  • MDL number MFCD03841948
  • Cas Number : 898792-50-2
  • Molecular Weight : 347.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0108-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0108-00
Availability details for
Quantity
  • Availability
  • Add to Cart