RIE156324734 Rieke Metals, Inc.

3-(piperidinomethyl)benzophenone

  • MDL number MFCD03841950
  • Cas Number : 898792-54-6
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-01
Availability details for
Quantity
  • Availability
  • Add to Cart