RIE156324735 Rieke Metals, Inc.

2-methyl-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841951
  • Cas Number : 898792-56-8
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methylphenyl)[3-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO/c1-16-8-3-4-11-19(16)20(22)18-10-7-9-17(14-18)15-21-12-5-2-6-13-21/h3-4,7-11,14H,2,5-6,12-13,15H2,1H3
Inchi Key KFGAYXMRSHKKLJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0200-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-02
Availability details for
Quantity
  • Availability
  • Add to Cart