RIE156324740 Rieke Metals, Inc.

4'-methoxy-3-piperidinomethyl benzophenone

  • MDL number MFCD03841956
  • Cas Number : 898792-66-0
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-07
Availability details for
Quantity
  • Availability
  • Add to Cart