RIE156324743 Rieke Metals, Inc.

4'-cyano-3-piperidinomethyl benzophenone

  • MDL number MFCD03841959
  • Cas Number : 898792-72-8
  • Molecular Weight : 304.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-10
Availability details for
Quantity
  • Availability
  • Add to Cart