RIE156324745 Rieke Metals, Inc.

3-carboethoxy-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841961
  • Cas Number : 898792-78-4
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-12
Availability details for
Quantity
  • Availability
  • Add to Cart