RIE156324746 Rieke Metals, Inc.

4'-carboethoxy-3-piperidinomethyl benzophenone

  • MDL number MFCD03841962
  • Cas Number : 898792-81-9
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[3-(1-piperidinylmethyl)benzoyl]benzoate
Inchi Code 1S/C22H25NO3/c1-2-26-22(25)19-11-9-18(10-12-19)21(24)20-8-6-7-17(15-20)16-23-13-4-3-5-14-23/h6-12,15H,2-5,13-14,16H2,1H3
Inchi Key XOIAUMJPZWFULQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0200-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-13
Availability details for
Quantity
  • Availability
  • Add to Cart