RIE156324747 Rieke Metals, Inc.

3'-piperidinomethyl-2-thiomethylbenzophenone

  • MDL number MFCD03841963
  • Cas Number : 898792-84-2
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-14
Availability details for
Quantity
  • Availability
  • Add to Cart