RIE156324748 Rieke Metals, Inc.

3-piperidinomethyl-4'-thiomethylbenzophenone

  • MDL number MFCD03841964
  • Cas Number : 898792-87-5
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-15
Availability details for
Quantity
  • Availability
  • Add to Cart