RIE156324752 Rieke Metals, Inc.

4'-chloro-3-piperidinomethyl benzophenone

  • MDL number MFCD03841968
  • Cas Number : 898792-99-9
  • Molecular Weight : 313.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-21
Availability details for
Quantity
  • Availability
  • Add to Cart