RIE156324753 Rieke Metals, Inc.

3-fluoro-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841969
  • Cas Number : 898793-02-7
  • Molecular Weight : 297.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluorophenyl)[3-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H20FNO/c20-18-9-5-8-17(13-18)19(22)16-7-4-6-15(12-16)14-21-10-2-1-3-11-21/h4-9,12-13H,1-3,10-11,14H2
Inchi Key DVHVOCYNZZGSIR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0200-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-22
Availability details for
Quantity
  • Availability
  • Add to Cart