RIE156324755 Rieke Metals, Inc.

2,3-dimethyl-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841971
  • Cas Number : 898793-08-3
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-26
Availability details for
Quantity
  • Availability
  • Add to Cart