RIE156324756 Rieke Metals, Inc.

2,4-dimethyl-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841972
  • Cas Number : 898793-11-8
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-27
Availability details for
Quantity
  • Availability
  • Add to Cart