RIE156324757 Rieke Metals, Inc.

2,5-dimethyl-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841973
  • Cas Number : 898793-14-1
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-28
Availability details for
Quantity
  • Availability
  • Add to Cart