RIE156324761 Rieke Metals, Inc.

4-bromo-3-fluoro-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841977
  • Cas Number : 898793-24-3
  • Molecular Weight : 376.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-32
Availability details for
Quantity
  • Availability
  • Add to Cart