RIE156324766 Rieke Metals, Inc.

3'-piperidinomethyl-2-trifluoromethylbenzophenone

  • MDL number MFCD03841982
  • Cas Number : 898793-34-5
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-piperidinylmethyl)phenyl][2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C20H20F3NO/c21-20(22,23)18-10-3-2-9-17(18)19(25)16-8-6-7-15(13-16)14-24-11-4-1-5-12-24/h2-3,6-10,13H,1,4-5,11-12,14H2
Inchi Key ROOUZWKSHJZISN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0200-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-43
Availability details for
Quantity
  • Availability
  • Add to Cart