RIE156324776 Rieke Metals, Inc.

3,4-dichloro-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841992
  • Cas Number : 898793-54-9
  • Molecular Weight : 348.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-56
Availability details for
Quantity
  • Availability
  • Add to Cart