RIE156324779 Rieke Metals, Inc.

3,4-difluoro-3'-piperidinomethyl benzophenone

  • MDL number MFCD03841995
  • Cas Number : 898793-60-7
  • Molecular Weight : 315.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0200-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-59
Availability details for
Quantity
  • Availability
  • Add to Cart