RIE156324781 Rieke Metals, Inc.

3'-piperidinomethyl-3,4,5-trifluorobenzophenone

  • MDL number MFCD03841997
  • Cas Number : 898793-64-1
  • Molecular Weight : 333.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-piperidinylmethyl)phenyl](3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C19H18F3NO/c20-16-10-15(11-17(21)18(16)22)19(24)14-6-4-5-13(9-14)12-23-7-2-1-3-8-23/h4-6,9-11H,1-3,7-8,12H2
Inchi Key VWWZMUSYYPHNGY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0200-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0200-61
Availability details for
Quantity
  • Availability
  • Add to Cart