RIE156324784 Rieke Metals, Inc.

cyclopentyl 3-(piperidinomethyl)phenyl ketone

  • MDL number MFCD03842000
  • Cas Number : 898793-70-9
  • Molecular Weight : 271.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0203-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0203-00
Availability details for
Quantity
  • Availability
  • Add to Cart