RIE156324786 Rieke Metals, Inc.

ethyl 4-oxo-4-[3-(piperidinomethyl)phenyl]butyrate

  • MDL number MFCD03842002
  • Cas Number : 898793-74-3
  • Molecular Weight : 303.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0205-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0205-00
Availability details for
Quantity
  • Availability
  • Add to Cart