RIE156324788 Rieke Metals, Inc.

ethyl 6-oxo-6-[3-(piperidinomethyl)phenyl]hexanoate

  • MDL number MFCD03842004
  • Cas Number : 898793-80-1
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-oxo-6-[3-(1-piperidinylmethyl)phenyl]hexanoate
Inchi Code 1S/C20H29NO3/c1-2-24-20(23)12-5-4-11-19(22)18-10-8-9-17(15-18)16-21-13-6-3-7-14-21/h8-10,15H,2-7,11-14,16H2,1H3
Inchi Key BKAKRYBRVKYGPD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0207-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0207-00
Availability details for
Quantity
  • Availability
  • Add to Cart