RIE156324793 Rieke Metals, Inc.

3-methyl-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842009
  • Cas Number : 898793-94-7
  • Molecular Weight : 279.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methylphenyl)[3-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H21NO/c1-15-6-4-8-17(12-15)19(21)18-9-5-7-16(13-18)14-20-10-2-3-11-20/h4-9,12-13H,2-3,10-11,14H2,1H3
Inchi Key GQSYNYJMCDVEOH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0300-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-03
Availability details for
Quantity
  • Availability
  • Add to Cart