RIE156324794 Rieke Metals, Inc.

4'-methyl-3-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842010
  • Cas Number : 898793-97-0
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-04
Availability details for
Quantity
  • Availability
  • Add to Cart