RIE156324796 Rieke Metals, Inc.

3-methoxy-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842012
  • Cas Number : 898794-03-1
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-06
Availability details for
Quantity
  • Availability
  • Add to Cart